Learn More
S2 Media 100mL Butterfield's Phosphate Buffer (Phosphate Buffer, pH 7.2) in 120mL HiClarity Flip-Top Tube, 1/each
Catalog No. 502995767
Encompass_Preferred
Butterfield's Phosphate Buffer (BPB), pH 7.2, is utilized as a diluent in the preparation of sample dilutions and microbiological suspensions in laboratory processes. • Product meets AOAC, APHA, ASM, BAM, SM-Dairy, SMWW, Harmonized USP/EP/JP, and USDA reference methods and performance criteria.• Butterfield's Phosphate Buffer should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Phosphate Buffer, pH 7.2, and Butterfield's Phosphate Buffered Dilution Water.
This item is not returnable. View return policy
Catalog No. 50-299-5767 Supplier S2 Media Supplier No. 5252
Butterfield's Phosphate Buffer (BPB), pH 7.2, is utilized as a diluent in the preparation of sample dilutions and microbiological suspensions in laboratory processes. • Product meets AOAC, APHA, ASM, BAM, SM-Dairy, SMWW, Harmonized USP/EP/JP, and USDA reference methods and performance criteria.• Butterfield's Phosphate Buffer should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Phosphate Buffer, pH 7.2, and Butterfield's Phosphate Buffered Dilution Water.