Learn More
Ace Glass, Inc. Extraction apparatus, liquid from ether, 100mL, complete with 6740-04 condenser, Por B funnel tube, outer chamber and 6965-22 Erlenmeyer flask
SDP
Catalog No. 50973449
Encompass_Preferred
100mL extraction apparatus designed for the extracting of liquids with ether. The dispersion of the ether is accomplished with the use of a 70-100micron porosity ACE fritted disc, which is attached to the base of the funnel tube.
Catalog No. 50-973-449 Supplier Ace Glass, Inc. Supplier No. 684030
100mL extraction apparatus designed for the extracting of liquids with ether. The dispersion of the ether is accomplished with the use of a 70-100micron porosity ACE fritted disc, which is attached to the base of the funnel tube.