Learn More
S2 Media 100mL Fluid Thioglycollate Medium (FTM) with Indicator in 125mL Media Bottle with Septum Cap, 1/each
Catalog No. 502995799
Encompass_Preferred
Fluid Thioglycollate Medium (FTM) with Indicator is a general purpose medium recommended for sterility testing and for the cultivation of anaerobes, aerobes, and microaerophiles. Casein and yeast extract provide nitrogen, essential vitamins, and amino acids. Glucose provides a carbon source, sodium chloride maintains osmotic balance, and L-cystine and sodium thioglycollate act as reducing agents. This is supported by a low amount of agar, which reduces the oxygen permeability through the medium. Resazurin is an oxidation indicator. Sodium thioglycollate inactivates mercurial compounds present in samples.• Product meets AOAC, APHA, BAM, CLSI, Harmonized USP/EP/JP, and ISO reference methods and performance criteria.• FTM should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Septum Cap on Plastic Bottle
This item is not returnable. View return policy
Catalog No. 50-299-5799 Supplier S2 Media Supplier No. 5432
Fluid Thioglycollate Medium (FTM) with Indicator is a general purpose medium recommended for sterility testing and for the cultivation of anaerobes, aerobes, and microaerophiles. Casein and yeast extract provide nitrogen, essential vitamins, and amino acids. Glucose provides a carbon source, sodium chloride maintains osmotic balance, and L-cystine and sodium thioglycollate act as reducing agents. This is supported by a low amount of agar, which reduces the oxygen permeability through the medium. Resazurin is an oxidation indicator. Sodium thioglycollate inactivates mercurial compounds present in samples.• Product meets AOAC, APHA, BAM, CLSI, Harmonized USP/EP/JP, and ISO reference methods and performance criteria.• FTM should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Septum Cap on Plastic Bottle