Learn More
Microsolv Technology Corporation Bottle, HPLC Mobile Phase Storage, Glass, Amber, 100ml, Screw Top, GL45 Threads. 1 EA.
Catalog No. 502269793
Encompass_Preferred
This Amber, Borosilicate Glass Bottle has a 100ml "Total Fill Capacity" and is perfect as an HPLC Mobile Phase Storage Bottle. The small opening and Enamel Graduations and Marking Patch makes this a convenient way to store concentrated Mobile Phase or its Ingredients. A Blue, Solid Screw Cap without a liner and a polypropylene Drip Ring is included. This Bottle is Not "Safety Coated".
This item is not returnable. View return policy
Catalog No. 50-226-9793 Supplier Microsolv Technology Corporation Supplier No. 58807100A
This Amber, Borosilicate Glass Bottle has a 100ml "Total Fill Capacity" and is perfect as an HPLC Mobile Phase Storage Bottle. The small opening and Enamel Graduations and Marking Patch makes this a convenient way to store concentrated Mobile Phase or its Ingredients. A Blue, Solid Screw Cap without a liner and a polypropylene Drip Ring is included. This Bottle is Not "Safety Coated".