Learn More
Microsolv Technology Corporation Bottles, HPLC Mobile Phase Storage, Glass, Amber, 100ml, Screw Top, GL45 Threads. 10/CS.
Catalog No. 502269794
Encompass_Preferred
These Amber, Borosilicate Glass Bottles have a 100ml "Total Fill Capacity" and are perfect as HPLC Mobile Phase Storage Bottles. The small openings and Enamel Graduations and Marking Patches makes them a convenient way to store concentrated Mobile Phase or its Ingredients.Blue, Solid Screw Caps without liners and polypropylene Drip Rings are included. This Bottle is Not "Safety Coated".
This item is not returnable. View return policy
Catalog No. 50-226-9794 Supplier Microsolv Technology Corporation Supplier No. 58807100ACS
These Amber, Borosilicate Glass Bottles have a 100ml "Total Fill Capacity" and are perfect as HPLC Mobile Phase Storage Bottles. The small openings and Enamel Graduations and Marking Patches makes them a convenient way to store concentrated Mobile Phase or its Ingredients.Blue, Solid Screw Caps without liners and polypropylene Drip Rings are included. This Bottle is Not "Safety Coated".