Learn More
CELLTREAT Scientific Products 100mL Graduated Beaker With Handle, Polypropylene, Non-sterile, Case Quantity 12
SDP
Catalog No. 5000670046
Encompass_Preferred
Molded pouring spouts and rolled lip fordripless work environment
Handled Beakers feature clearly printedgraduations
Sturdy handles with textured thumb restensure safe handling of hazardous solutions
Polypropylene material is resistant to avariety of strong acids, bases, and solvents
Can withstand -20 ̊C to 135 ̊C
Catalog No. 50-006-70046 Supplier CELLTREAT Scientific Products Supplier No. 230521
Add to Cart
Add to Cart
Molded pouring spouts and rolled lip fordripless work environment
Handled Beakers feature clearly printedgraduations
Sturdy handles with textured thumb restensure safe handling of hazardous solutions
Polypropylene material is resistant to avariety of strong acids, bases, and solvents
Can withstand -20 ̊C to 135 ̊C