Learn More
CELLTREAT Scientific Products 100mL Graduated Cylinder, Polymethylpentene, Non-sterile, Case Quantity 4
Supplier: CELLTREAT Scientific Products 230534
Graduated Cylinders are designed with ring-molded, incrementalgraduations to ensure excellent readability and accuracy
Premium polymethylpentene material is a transparent,lightweight, and shatterproof alternative to glass measuringcylinders, with strong chemical resistance and a maximumtemperature tolerance of 135 ̊C
Pour spout and rolled lip design prevent drips and splashesCylinders feature a wide hexagonal base for increased stability