Learn More
Chemglass Life Sciences 100mL Pear Shaped Flask, 19/22 Outer Joint
SDP
Catalog No. 50873172
Encompass_Preferred
100mL Pear Shaped Flask, 19/22 Outer Joint. Pear shape HEAVY WALL flask with a standard taper outer joint.
Catalog No. 50-873-172 Supplier Chemglass Life Sciences Supplier No. CG155431
100mL Pear Shaped Flask, 19/22 Outer Joint. Pear shape HEAVY WALL flask with a standard taper outer joint.