Learn More
Ace Glass, Inc. 100mL graduated funnel, #25 Ace-Thred bottom
SDP
Catalog No. 50978185
Encompass_Preferred
100mL graduated funnel with a #25 Ace-Thred bottom. Funnel is part of the ACE borosilicate glass filtration apparatus that accomm ODates 25mm membrane filters.
Catalog No. 50-978-185 Supplier Ace Glass, Inc. Supplier No. 370002
100mL graduated funnel with a #25 Ace-Thred bottom. Funnel is part of the ACE borosilicate glass filtration apparatus that accomm ODates 25mm membrane filters.