Learn More
Chemglass Life Sciences 100mL Pear Shaped Flask, 14/20 Outer Joint
SDP
Catalog No. 501215264
Encompass_Preferred
100mL Pear Shaped Flask, 14/20 Outer Joint. Pear shape HEAVY WALL flask with a standard taper outer joint.
Catalog No. 50-121-5264 Supplier Chemglass Life Sciences Supplier No. CG155430
100mL Pear Shaped Flask, 14/20 Outer Joint. Pear shape HEAVY WALL flask with a standard taper outer joint.