Learn More
S2 Media 100mL 1x Phosphate Buffered Saline (PBS), pH 7.5, 125mL PP Boston Round Bottle, 1ea
Catalog No. 502995787
Encompass_Preferred
1x Phosphate Buffered Saline (PBS), pH 7.5 has many applications due to its isotonic nature, including uses as a rinse fluid, diluent, and buffer. Phosphate salts are nontoxic to living cells and have a high buffering capacity. pH maintenance is important in retaining cell viability and in the recovery and revitalization of injured or damaged cells during bacterial culture and analysis. PBS is generally used as a 10mM working solution containing a mixture of sodium chloride and inorganic phosphate salts.• Product meets AOAC, APHA, BAM, CLSI, SM-Dairy, SMWW, and USP-NF reference methods and performance criteria.• For in vitro diagnostic (IVD) use.• PBS should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5787 Supplier S2 Media Supplier No. 5472
Add to Cart
Add to Cart
1x Phosphate Buffered Saline (PBS), pH 7.5 has many applications due to its isotonic nature, including uses as a rinse fluid, diluent, and buffer. Phosphate salts are nontoxic to living cells and have a high buffering capacity. pH maintenance is important in retaining cell viability and in the recovery and revitalization of injured or damaged cells during bacterial culture and analysis. PBS is generally used as a 10mM working solution containing a mixture of sodium chloride and inorganic phosphate salts.• Product meets AOAC, APHA, BAM, CLSI, SM-Dairy, SMWW, and USP-NF reference methods and performance criteria.• For in vitro diagnostic (IVD) use.• PBS should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.