Learn More
S2 Media 100mL Potato Dextrose Broth (PDB) in 125mL Media Bottle, 1/each
Catalog No. 502995920
Encompass_Preferred
10mL Potato Dextrose Broth (PDA) is an enrichment broth used for the cultivation of yeasts and molds. The low pH of this medium can inhibit some bacterial growth. • Product meets AOAC, APHA, ASM, CLSI, Harmonized USP/EP/JP, and SM-Dairy reference methods and performance criteria.• PDB should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5920 Supplier S2 Media Supplier No. 5300B
10mL Potato Dextrose Broth (PDA) is an enrichment broth used for the cultivation of yeasts and molds. The low pH of this medium can inhibit some bacterial growth. • Product meets AOAC, APHA, ASM, CLSI, Harmonized USP/EP/JP, and SM-Dairy reference methods and performance criteria.• PDB should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.