Learn More
Ace Glass, Inc. 100mL side-operated, screw-feed powder dispensing funnel
SDP
Catalog No. 50974719
Encompass_Preferred
100mL side-operated, screw-feed powder dispensing funnel, for use with up to 25 mesh. Features a #15 Ace-Thred at the top, a large, 1-1/4 knob, and top female and bottom male 24/40 joints.
Catalog No. 50-974-719 Supplier Ace Glass, Inc. Supplier No. 723930
Add to Cart
Add to Cart
100mL side-operated, screw-feed powder dispensing funnel, for use with up to 25 mesh. Features a #15 Ace-Thred at the top, a large, 1-1/4 knob, and top female and bottom male 24/40 joints.