Learn More
Beckman Coulter 100ML PP CENTTUBES BX/6 38X102
Catalog No. NC9881759
Encompass_Preferred
Tubes, Polypropylene, 100 mL, Size: 38 x 102 mm, 6/BX
Catalog No. NC9881759 Supplier Beckman Coulter Supplier No. 355624
Tubes, Polypropylene, 100 mL, Size: 38 x 102 mm, 6/BX