Learn More
Chemglass Life Sciences 100mL 2-Neck Round Bottom Flask (RBF) , 1-CN 24/40 Outer, 1-SN 24/40 Outer
SDP
Catalog No. CG151801
Encompass_Preferred
100mL 2-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 1-SN 24/40 Outer. Round bottom, HEAVY WALL two neck flask with standard taper outer joints on both the center neck and vertical side neck.
Catalog No. CG151801 Supplier Chemglass Life Sciences Supplier No. CG151801
100mL 2-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 1-SN 24/40 Outer. Round bottom, HEAVY WALL two neck flask with standard taper outer joints on both the center neck and vertical side neck.