Learn More
Chemglass Life Sciences Flask, Round Bottom, 100mL, Heavy Wall, 3-Neck, 19/22 CN - 14/20 SN's Angled 20°
SDP
Catalog No. CG1524A09
Encompass_Preferred
Flask, Round Bottom, 100mL, Heavy Wall, 19/22 - 14/20, 3-Neck, Angled 20¿.Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 19/22Side neck: 14/20
Catalog No. CG1524A09 Supplier Chemglass Life Sciences Supplier No. CG1524A09
Flask, Round Bottom, 100mL, Heavy Wall, 19/22 - 14/20, 3-Neck, Angled 20¿.Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 19/22Side neck: 14/20