Learn More
Chemglass Life Sciences 100mL Single Neck Round Bottom Flask (RBF) , 35/25 Socket Joint
SDP
Catalog No. CG150804
Encompass_Preferred
100mL Single Neck Round Bottomed Flask 35/25 Socket Joint. Round bottom, HEAVY WALL single short neck flask with a spherical socket joint.
Catalog No. CG150804 Supplier Chemglass Life Sciences Supplier No. CG150804
100mL Single Neck Round Bottomed Flask 35/25 Socket Joint. Round bottom, HEAVY WALL single short neck flask with a spherical socket joint.