Learn More
Andwin Scientific 100 mL Reservoir, Case of 100
SDP
Catalog No. 50930352
Encompass_Preferred
Non Sterile disposable modified polystyrene reservoirs for use with multichannel pipettors. Capacity: 100mL. White color. 1/bag.
Catalog No. 50-930-352 Supplier Andwin Scientific Supplier No. 7049710
Non Sterile disposable modified polystyrene reservoirs for use with multichannel pipettors. Capacity: 100mL. White color. 1/bag.