Learn More
CELLTREAT Scientific Products 100mL Reagent Reservoir, Individually Wrapped, Sterile
SDP
Catalog No. NC1772278
Encompass_Preferred
CELLTREAT Scientific Products Polystyrene Reagent Reservoirs are designed to assist in repetitive liquid transfer. White color enhances solution visibility. Angled design promotes funneling of liquid to maximize retrieval, and works with multiple-channel pipettes. Pour spouts on corners and textured gripping areas on the side. Low profile is ideal for small volume robotic tips. Bulk packed reservoirs are easy to separate. Available in a wide range of sizes for all of your pipetting needs.
Catalog No. NC1772278 Supplier CELLTREAT Scientific Products Supplier No. 229298
CELLTREAT Scientific Products Polystyrene Reagent Reservoirs are designed to assist in repetitive liquid transfer. White color enhances solution visibility. Angled design promotes funneling of liquid to maximize retrieval, and works with multiple-channel pipettes. Pour spouts on corners and textured gripping areas on the side. Low profile is ideal for small volume robotic tips. Bulk packed reservoirs are easy to separate. Available in a wide range of sizes for all of your pipetting needs.