Learn More
Bellco Glass, Inc. 100mL Reagent Reservoir Cooler with Lid, Sterile
SDP
Catalog No. 502330601
Encompass_Preferred
Reusable reagent reservoir cooler is a custom-fit companion for 100 mL reagent reservoirsManufactured with non-toxic gel encased in heavy duty PVCBright blue coolers chill or freeze quickly and maintain reagents at <5°C for up to 1 hour
Catalog No. 50-233-0601 Supplier Bellco Glass, Inc. Supplier No. 451900237
Reusable reagent reservoir cooler is a custom-fit companion for 100 mL reagent reservoirsManufactured with non-toxic gel encased in heavy duty PVCBright blue coolers chill or freeze quickly and maintain reagents at <5°C for up to 1 hour