Learn More
Ace Glass, Inc. 100mL graduated Schlenk storage tube, 14/20 outer top, 14/35 inner side, stopcock inlet
SDP
Catalog No. 50975379
Encompass_Preferred
Catalog No. 50-975-379 Supplier Ace Glass, Inc. Supplier No. 775617