Learn More
S2 Media 100mL Sabouraud Dextrose Broth with Penicillin G (SDB-P), in 125mL PET/PETG Media Bottle, 1/each
Catalog No. 502995745
Encompass_Preferred
Sabouraud Dextrose Broth with Penicillin G (SDB-P) is a selective broth used in qualitative procedures for the cultivation of fungi. Emmons modified the formulation of Sabouraud by reducing the amount of dextrose from 40g/L to 20g/L and removing agar. This product is also referred to as Sabouraud Dextrose Broth 2%, due to the reduced dextrose content. Reference the appropriate standard method for additional details on test instructions. The broth is supplemented with 0.2µg/mL Penicillin G to inhibit gram-positive bacteria.• Product meets BAM, CLSI, ISO, NF, and Harmonized USP/EP/JP reference methods and performance criteria.• SDB-P should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5745 Supplier S2 Media Supplier No. 5087
Sabouraud Dextrose Broth with Penicillin G (SDB-P) is a selective broth used in qualitative procedures for the cultivation of fungi. Emmons modified the formulation of Sabouraud by reducing the amount of dextrose from 40g/L to 20g/L and removing agar. This product is also referred to as Sabouraud Dextrose Broth 2%, due to the reduced dextrose content. Reference the appropriate standard method for additional details on test instructions. The broth is supplemented with 0.2µg/mL Penicillin G to inhibit gram-positive bacteria.• Product meets BAM, CLSI, ISO, NF, and Harmonized USP/EP/JP reference methods and performance criteria.• SDB-P should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.