Learn More
S2 Media 100mL Sabouraud Dextrose Broth (SabDex Broth, SDB) in 125mL Media Bottle, 1ea
Catalog No. 502995769
Encompass_Preferred
Sabouraud Dextrose Broth (low pH) is a selective and non-differential culture medium, which can be used as an enrichment broth. SabDex Broth is used for the cultivation of fungi and aciduric microorganisms. It is recommended for the cultivation of fungi from sterile and non-sterile products by the Harmonized Pharmacopoeia. • Product meets AOAC, APHA, ASM, BAM, CDC, CLSI, Harmonized USP/EP/JP, and SM-Dairy reference methods and performance criteria.• SabDex Broth should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Fluid Sabouraud Medium, Sabouraud Dextrose Broth (Low pH), and Sabouraud Dextrose Broth 2%• Septum caps available - contact S2Media (info@s2cm.com)
This item is not returnable. View return policy
Catalog No. 50-299-5769 Supplier S2 Media Supplier No. 5262
Sabouraud Dextrose Broth (low pH) is a selective and non-differential culture medium, which can be used as an enrichment broth. SabDex Broth is used for the cultivation of fungi and aciduric microorganisms. It is recommended for the cultivation of fungi from sterile and non-sterile products by the Harmonized Pharmacopoeia. • Product meets AOAC, APHA, ASM, BAM, CDC, CLSI, Harmonized USP/EP/JP, and SM-Dairy reference methods and performance criteria.• SabDex Broth should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Fluid Sabouraud Medium, Sabouraud Dextrose Broth (Low pH), and Sabouraud Dextrose Broth 2%• Septum caps available - contact S2Media (info@s2cm.com)