Learn More
MOLDPRO INC 100 mL Titration Cup, 53.6 x 90.5 mm, Clear, 450/case, Flat Bottom, Graduated, PP Homopolymer
Catalog No. NC2233823
Encompass_Preferred
MoldPro MP-118 100 mL beaker is widely used size for titration/TAN/TBN systems by Mettler-Toledo (M-T). Comparable to M-T 00101974, 511093388 & 51109389; Dyn-a-med 80099; Precision Weighing Balances 99008; Fisher Scientific NC9485544; VWR 56221-531 & 12578-108. • 450 beakers/case • Made from virgin polypropylene • Graduations at 20, 40, 60, 80 and 100 mL • Excellent visibility • Good resistance to acids Fits the following M-T sample introduction systems: • Rondolino™ • Rondo™ • InMotion™ Flex Autosampler • InMotion™ Pro Autosampler • InMotion™ Max Autosampler Used on the following M-T instruments: • G10S • G20S • T5 • T7• T9
This item is not returnable. View return policy
Catalog No. NC2233823 Supplier MOLDPRO INC Supplier No. MP118
MoldPro MP-118 100 mL beaker is widely used size for titration/TAN/TBN systems by Mettler-Toledo (M-T). Comparable to M-T 00101974, 511093388 & 51109389; Dyn-a-med 80099; Precision Weighing Balances 99008; Fisher Scientific NC9485544; VWR 56221-531 & 12578-108. • 450 beakers/case • Made from virgin polypropylene • Graduations at 20, 40, 60, 80 and 100 mL • Excellent visibility • Good resistance to acids Fits the following M-T sample introduction systems: • Rondolino™ • Rondo™ • InMotion™ Flex Autosampler • InMotion™ Pro Autosampler • InMotion™ Max Autosampler Used on the following M-T instruments: • G10S • G20S • T5 • T7• T9