Learn More
Ace Glass, Inc. Scoop, 100mL, polypropylene, 102mm long handle, 92x48x35mm bowl, pk/12
SDP
Supplier:  Ace Glass, Inc. 1339507
Encompass
Scoop, 100mL, polypropylene, 102mm long handle, 92x48x35mm bowl, pk/12
Catalog No. NC1980862