Learn More
CELLTREAT Scientific Products 100mL Pipet, Individually Wrapped, Plastic/Plastic, Bag, Sterile, 50/CS
SDP
Catalog No. 501535143
Encompass_Preferred
CELLTREAT Scientific Products 100mL Serological Pipet in Plastic/Plastic Wrapper. Peel-to-open plastic/plastic wrappers are the ideal option for eliminating particulate; proven benefit over traditional paper/plastic wrapper. Wrappers reduce static cling versus other brands of plastic wrap pipets. All plastic wrapper can be wiped with alcohol without smearing ink or penetrating paper. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. 100mL pipet is ideal for large volume pipetting. Manufactured from high clarity virgin polysytrene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10 Individually Wrapped/bag, 50/case.
Catalog No. 50-153-5143 Supplier CELLTREAT Scientific Products Supplier No. 229220A
CELLTREAT Scientific Products 100mL Serological Pipet in Plastic/Plastic Wrapper. Peel-to-open plastic/plastic wrappers are the ideal option for eliminating particulate; proven benefit over traditional paper/plastic wrapper. Wrappers reduce static cling versus other brands of plastic wrap pipets. All plastic wrapper can be wiped with alcohol without smearing ink or penetrating paper. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. 100mL pipet is ideal for large volume pipetting. Manufactured from high clarity virgin polysytrene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10 Individually Wrapped/bag, 50/case.