Learn More
Chemglass Life Sciences 100mL Storage Flask, Cajon, Airfree, Schlenk, 0-4mm Valve, 135mm OAH
SDP
Catalog No. 50873069
Encompass_Preferred
100mL Storage Flask, Cajon, Airfree, Schlenk, 0-4mm Valve, 135mm OAH. Storage flask sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.
Catalog No. 50-873-069 Supplier Chemglass Life Sciences Supplier No. AF009402
100mL Storage Flask, Cajon, Airfree, Schlenk, 0-4mm Valve, 135mm OAH. Storage flask sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.