Learn More
S2 Media 100mL Tryptic Soy Broth with Penicillin G (TSB-P) / Soybean-Casein Digest (SCD) Medium with Penicillin G, in 125mL PET/PETG Media Bottle, 1/each
Catalog No. 502995812
Encompass_Preferred
Tryptic Soy Broth with Penicillin G (TSB-P) is a general nutrient and enrichment broth, supplemented to be selective. It is used in qualitative procedures for the cultivation of a wide variety of microorganisms. TSB is also recommended for sterility testing. Reference the appropriate standard method for additional details on test instructions. The broth is supplemented with 0.2µg/mL Penicillin G to inhibit gram-positive bacteria.• Product meets AOAC, BAM, CLSI, EPA, ISO, NF, SM-Dairy, SMWW, USDA, and Harmonized USP/EP/JP reference methods and performance criteria.• TSB-P should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5812 Supplier S2 Media Supplier No. 5088
Tryptic Soy Broth with Penicillin G (TSB-P) is a general nutrient and enrichment broth, supplemented to be selective. It is used in qualitative procedures for the cultivation of a wide variety of microorganisms. TSB is also recommended for sterility testing. Reference the appropriate standard method for additional details on test instructions. The broth is supplemented with 0.2µg/mL Penicillin G to inhibit gram-positive bacteria.• Product meets AOAC, BAM, CLSI, EPA, ISO, NF, SM-Dairy, SMWW, USDA, and Harmonized USP/EP/JP reference methods and performance criteria.• TSB-P should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.