Learn More
Ace Glass, Inc. 100mL vessel with sloped shoulders and a #25 Ace-Thred neck
SDP
Catalog No. 50973321
Encompass_Preferred
100mL vessel with sloped shoulders and a #25 Ace-Thred neck; includes PTFE lined cap. For use in freeze drying and storage. After drying material, vessel can be capped and stored. Working capacity of vessel is about 1/2 of volume.
Catalog No. 50-973-321 Supplier Ace Glass, Inc. Supplier No. 669620
100mL vessel with sloped shoulders and a #25 Ace-Thred neck; includes PTFE lined cap. For use in freeze drying and storage. After drying material, vessel can be capped and stored. Working capacity of vessel is about 1/2 of volume.