Learn More
Bellco Glass, Inc. 100mL Wobble-not Serological Pipet, Individually Wrapped, Sterile
SDP
Catalog No. 502329406
Encompass_Preferred
Wobble-not serological pipets have a patented innovative 2-tier step design at the plug end vs. traditional pipets that have a straight cylindrical-shaped plug end, requiring less insertion force.Wobble-nots are the only serological pipets that will give you two zones of contact in your pipet controller’s nozzle, to increase stability and reduce dripping.Compatible with any pipet controller.Ultra-clear markings and color-coded stripes for quick volume identification.25 & 50mL pipets feature negative graduations to -10mL.Premium polymer fiber barrier plug.Accuracy ± 2% at full volume.Pack Description – 10 Individually Wrapped/BagCase Quantity – 50Sterile – YesNon-pyrogenic – YesPrimary Color – ClearSecondary Color – PinkPrimary Material – PolystyreneVolume – 100mLGraduated – YesGraduation Increments (Subdivisions) – 1mLNegative Graduations – NARNase/DNase Free – Yes
Catalog No. 50-232-9406 Supplier Bellco Glass, Inc. Supplier No. 229095B00
Wobble-not serological pipets have a patented innovative 2-tier step design at the plug end vs. traditional pipets that have a straight cylindrical-shaped plug end, requiring less insertion force.Wobble-nots are the only serological pipets that will give you two zones of contact in your pipet controller’s nozzle, to increase stability and reduce dripping.Compatible with any pipet controller.Ultra-clear markings and color-coded stripes for quick volume identification.25 & 50mL pipets feature negative graduations to -10mL.Premium polymer fiber barrier plug.Accuracy ± 2% at full volume.Pack Description – 10 Individually Wrapped/BagCase Quantity – 50Sterile – YesNon-pyrogenic – YesPrimary Color – ClearSecondary Color – PinkPrimary Material – PolystyreneVolume – 100mLGraduated – YesGraduation Increments (Subdivisions) – 1mLNegative Graduations – NARNase/DNase Free – Yes