Learn More
Bellco Glass, Inc. 100mL Wobble-not Serological Pipet, Bulk Packed in Bags, Sterile
SDP
Catalog No. 502330534
Encompass_Preferred
Less Dripping with Wobble-not Wobble-not serological pipets have a patented innovative 2-tier step design at the plug end vs. traditional pipets that have a straight cylindrical-shaped plug end. Wobble-nots are the only serological pipets that will give you two zones of contact in your pipet controller's nozzle. Insert with less force. No jamming. Plus, you get maximum stability which means less dripping, more accuracy, and less risk of contamination!Low-Insertion Force Serological Pipet:Unique patented 2-tier step design at plug end3 packaging options, all Sterile: Individually wrapped in paper/plastic or plastic/plastic and bulk (zip lock resealable plastic bags)Sterile – RNase/DNase freeNon-pyrogenicAccuracy ± 2% at full volumeFilter at plug end made of porous polypropyleneUltra-clear markings
Catalog No. 50-233-0534 Supplier Bellco Glass, Inc. Supplier No. 451900163
Less Dripping with Wobble-not Wobble-not serological pipets have a patented innovative 2-tier step design at the plug end vs. traditional pipets that have a straight cylindrical-shaped plug end. Wobble-nots are the only serological pipets that will give you two zones of contact in your pipet controller's nozzle. Insert with less force. No jamming. Plus, you get maximum stability which means less dripping, more accuracy, and less risk of contamination!Low-Insertion Force Serological Pipet:Unique patented 2-tier step design at plug end3 packaging options, all Sterile: Individually wrapped in paper/plastic or plastic/plastic and bulk (zip lock resealable plastic bags)Sterile – RNase/DNase freeNon-pyrogenicAccuracy ± 2% at full volumeFilter at plug end made of porous polypropyleneUltra-clear markings