Learn More
Technical Glass Products Inc Clear Fused Quartz Crucible 100ML Capcity
SDP
Catalog No. NC1216667
Encompass
Clear Fused Quartz Crucible /100cc capacity, 62mm Top Dia x 66mm Tall, 35mm base diameter / Lid Sold Seperately
Catalog No. NC1216667 Supplier Technical Glass Products Inc Supplier No. FQCL100NL
Clear Fused Quartz Crucible /100cc capacity, 62mm Top Dia x 66mm Tall, 35mm base diameter / Lid Sold Seperately