Learn More
Chemglass Life Sciences Flask, Round Bottom, 100mL, Heavy Wall, 3-Neck, 19/22 CN - 19/22 SN's Angled 20°, Morton Indents
SDP
Catalog No. CG1525A04
Encompass_Preferred
Flask, Round Bottom, 100mL, Heavy Wall, 19/22 - 19/22, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center and 20¿ angled side necks. Indentations increase agitation of the contents of the flask during stirring.Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum
Catalog No. CG1525A04 Supplier Chemglass Life Sciences Supplier No. CG1525A04
Flask, Round Bottom, 100mL, Heavy Wall, 19/22 - 19/22, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center and 20¿ angled side necks. Indentations increase agitation of the contents of the flask during stirring.Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum