Learn More
Ace Glass, Inc. 100mm powder funnel with 24/29 inner joint
SDP
Catalog No. 50974712
Encompass_Preferred
100mm powder funnel with 24/29 inner joint is useful in pouring powders or liquids into ground joint containers.
Catalog No. 50-974-712 Supplier Ace Glass, Inc. Supplier No. 7236124
100mm powder funnel with 24/29 inner joint is useful in pouring powders or liquids into ground joint containers.