Learn More
Ace Glass, Inc. 100mm powder funnel with 29/32 inner joint
SDP
Catalog No. 50974713
Encompass_Preferred
100mm powder funnel with 29/32 inner joint is useful in pouring powders or liquids into ground joint containers.
Catalog No. 50-974-713 Supplier Ace Glass, Inc. Supplier No. 7236129
100mm powder funnel with 29/32 inner joint is useful in pouring powders or liquids into ground joint containers.