Learn More
Ace Glass, Inc. 100mm powder funnel with 35/25 inner joint
SDP
Catalog No. 50979225
Encompass_Preferred
100mm powder funnel with 35/25 inner joint is useful in pouring powders or liquids into ground joint containers.
Catalog No. 50-979-225 Supplier Ace Glass, Inc. Supplier No. 723620
Add to Cart
Add to Cart
100mm powder funnel with 35/25 inner joint is useful in pouring powders or liquids into ground joint containers.