Learn More
Chemglass Life Sciences Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 2-24/40 Side Necks, 1-10/30 Thermometer Joint Side Neck, 100mm Schott Flange
SDP
Catalog No. CG194201
Encompass_Preferred
Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 2-24/40 Side Necks, 1-10/30 Thermometer Joint Side Neck, 100mm Schott Flange. Four neck vessel lid to fit CG-1920 through CG-1936 vessels. The 10/30 outer joint will permit a thermometer to be inserted vertically to the bottom of the vessel. Lid has a flat ground surface allowing it to be greased or used with an o-ring.
Catalog No. CG194201 Supplier Chemglass Life Sciences Supplier No. CG194201
Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 2-24/40 Side Necks, 1-10/30 Thermometer Joint Side Neck, 100mm Schott Flange. Four neck vessel lid to fit CG-1920 through CG-1936 vessels. The 10/30 outer joint will permit a thermometer to be inserted vertically to the bottom of the vessel. Lid has a flat ground surface allowing it to be greased or used with an o-ring.