Learn More
Chemglass Life Sciences Flange, Compatible with Buchi Rotary Evaporators, Flange OD: 100mm, Flange ID: 74mm, Shank OD: 85mm
SDP
Catalog No. CG14602
Encompass_Preferred
Replacement flange compatible with Buchi condensers and rotary evaporator flasks. Flange OD: 100mm, Flange ID: 74mm, Shank OD: 85mm.
Catalog No. CG14602 Supplier Chemglass Life Sciences Supplier No. CG14602
Replacement flange compatible with Buchi condensers and rotary evaporator flasks. Flange OD: 100mm, Flange ID: 74mm, Shank OD: 85mm.