Learn More
Chemglass Life Sciences 100mm Cell Culture Dish, TC, Sterile
SDP
Supplier:  Chemglass Life Sciences CGE1800105
Encompass
100mm Cell Culture Dish, TC, Sterile. Cell culture dishes are ideal for all types of culturing where microscopic observation is required. Numeric marks on the bottom of each dish allow users to identify the location of cells.
Catalog No. 50-189-7568