Learn More
Chemglass Life Sciences Flange, Reaction Kettle, 100mm I.D., With O-Ring Groove
SDP
Catalog No. CG14702
Encompass_Preferred
Flange, Reaction Kettle, 100mm I.D., With O-Ring Groove. Flange is formed with a groove in the flat portion of the mating surface to accept an O-Ring. The flange also has a ground surface, permitting assembly with either the CG-148 flat flange or CG-149 lid blank using grease. In the greased application, the O-Ring groove prevents grease from entering the reaction kettle and causing possible contamination. Outer taper of flange is formed at a 45º angle to accept the CG-141 clamp.
Catalog No. CG14702 Supplier Chemglass Life Sciences Supplier No. CG14702
Flange, Reaction Kettle, 100mm I.D., With O-Ring Groove. Flange is formed with a groove in the flat portion of the mating surface to accept an O-Ring. The flange also has a ground surface, permitting assembly with either the CG-148 flat flange or CG-149 lid blank using grease. In the greased application, the O-Ring groove prevents grease from entering the reaction kettle and causing possible contamination. Outer taper of flange is formed at a 45º angle to accept the CG-141 clamp.