Learn More
Chemglass Life Sciences Reaction Kettle Flange, Duran, Flat Ground, Flange ID: 100mm
SDP
Catalog No. CG14802
Encompass_Preferred
Reaction Kettle Flange, Duran, Flat Ground, Flange ID: 100mm. Similar to CG-147, but formed without the o-ring groove.
Catalog No. CG14802 Supplier Chemglass Life Sciences Supplier No. CG14802
Reaction Kettle Flange, Duran, Flat Ground, Flange ID: 100mm. Similar to CG-147, but formed without the o-ring groove.