Learn More
Azzota Corp 100mm fluor cuvette 35ml glass
Catalog No. 5000887220
Encompass_Preferred
100mm Pathlength Standard Fluorometer Cuvettes 35ml Optical Glass (340nm-2500nm)
Catalog No. 50-008-87220 Supplier Azzota Corp Supplier No. G209
100mm Pathlength Standard Fluorometer Cuvettes 35ml Optical Glass (340nm-2500nm)