Learn More
Azzota Corp 100mm Fluor Cuvette 35ml Qz
Catalog No. 5000887166
Encompass_Preferred
100mm Pathlength Standard Fluorometer Cuvettes 35ml Quartz (190nm-2500nm)
Catalog No. 50-008-87166 Supplier Azzota Corp Supplier No. Q209
100mm Pathlength Standard Fluorometer Cuvettes 35ml Quartz (190nm-2500nm)