Learn More
Chemglass Life Sciences 100 mm O.D. x 50 mm I.D. Cork Flask Support Ring for Round-Bottom Flasks
SDP
Supplier:  Chemglass Life Sciences CG3060100
Encompass_Preferred
Cork flask support ring designed to cradle and stabilize round-bottom flasks. The concave top provides a secure seat for spherical glassware during storage, handling, or short-term support on benches.
  • Concave recess supports round-bottom flasks.
  • Outer diameter 100 mm, inner diameter 50 mm.
  • Made from natural cork for cushioning and grip.
  • Fits flasks approximately 150-250 mL.
  • Supplied individually (each).
Catalog No. NC2951657