Learn More
CELLTREAT Scientific Products 100mm x 15mm Petri Dish w/Grip Ring, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229693
Encompass_Preferred
CELLTREAT Scientific Products 100mm x 15mm Non-Treated Petri Dish. Dishes have thicker and more durable plastic than other Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 15-16mL and growth area of 55cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 500/case.
Catalog No. 50-202-038