Learn More
CELLTREAT Scientific Products 100mm x 10mm Petri Dish, Sterile
SDP
Catalog No. 5000144334
Encompass_Preferred
CELLTREAT Petri Dishes are manufactured in high capacity automated production lines resulting in high quality, value priced productsLow profile 10mm height saves space in limited cold storage areasVentilation ribs reduce condensation and allow air circulationConsistent flatness allows for even media distributionStackable - Lids contain stacking rings for increased stabilityDurable packaging offers greater protection from damage versus competitive brands
Catalog No. 50-001-44334 Supplier CELLTREAT Scientific Products Supplier No. 229691
CELLTREAT Petri Dishes are manufactured in high capacity automated production lines resulting in high quality, value priced productsLow profile 10mm height saves space in limited cold storage areasVentilation ribs reduce condensation and allow air circulationConsistent flatness allows for even media distributionStackable - Lids contain stacking rings for increased stabilityDurable packaging offers greater protection from damage versus competitive brands