Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bellco Glass, Inc. 100mm x 15mm Petri Dish
SDP

Catalog No. 502330222
Encompass_Preferred

Petri Dishes are manufactured in high capacity automated production lines resulting in high quality, value priced products. Ventilation ribs reduce condensation and allow air circulation.\n\nSpecifications:\n

    \n
  • Consistent flatness allows for even media distribution
  • \n
  • Stackable – Lids contain stacking rings for increased stability
  • \n
  • Stackable/Slideable – Features partial edge ring on base which allows a single dish to be slid from a large stack and enhances stability when stacked
  • \n
  • Slippable – Designed for automated filling and streaking systems, features side arrows and ISO Mark Target for automation
  • \n
  • Working Volume: 15-16mL
  • \n
  • Growth Area: 55cm2
  • \n
  • Case Quantity: 500
  • \n

Catalog No. 50-233-0222 Supplier Bellco Glass, Inc. Supplier No. 229695000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Petri Dishes are manufactured in high capacity automated production lines resulting in high quality, value priced products. Ventilation ribs reduce condensation and allow air circulation.\n\nSpecifications:\n

    \n
  • Consistent flatness allows for even media distribution
  • \n
  • Stackable – Lids contain stacking rings for increased stability
  • \n
  • Stackable/Slideable – Features partial edge ring on base which allows a single dish to be slid from a large stack and enhances stability when stacked
  • \n
  • Slippable – Designed for automated filling and streaking systems, features side arrows and ISO Mark Target for automation
  • \n
  • Working Volume: 15-16mL
  • \n
  • Growth Area: 55cm2
  • \n
  • Case Quantity: 500
  • \n

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.