Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bellco Glass, Inc. 100mm x 15mm Petri Dish
SDP

Catalog No. 502330311
Encompass_Preferred

Cell Culture Dishes are available in Tissue Culture Treated and Non-treated Cell Culture versions. Petri (Non-treated Cell Culture Dishes) have thicker and more durable plastic than standard Petri Dishes – both can be used for the same applications. 60, 70 and 100mm dishes available with a pronounced grip ring for improved handling.\n\nSpecifications:\n

    \n
  • Vented lids contain molded spacers to allow gas exchange (except Part No. 2296-55000 which has a non-vented lid)
  • \n
  • With griping feature
  • \n
  • Working Volume: 15-16mL
  • \n
  • Working Area: 55cm2
  • \n
  • Case Quantity: 300
  • \n

Catalog No. 50-233-0311 Supplier Bellco Glass, Inc. Supplier No. 229692000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell Culture Dishes are available in Tissue Culture Treated and Non-treated Cell Culture versions. Petri (Non-treated Cell Culture Dishes) have thicker and more durable plastic than standard Petri Dishes – both can be used for the same applications. 60, 70 and 100mm dishes available with a pronounced grip ring for improved handling.\n\nSpecifications:\n

    \n
  • Vented lids contain molded spacers to allow gas exchange (except Part No. 2296-55000 which has a non-vented lid)
  • \n
  • With griping feature
  • \n
  • Working Volume: 15-16mL
  • \n
  • Working Area: 55cm2
  • \n
  • Case Quantity: 300
  • \n

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.