Learn More
Bellco Glass, Inc. 100mm x 15mm Petri Dish
SDP
Catalog No. 502330346
Encompass_Preferred
CELLTREAT Petri Dishes are manufactured in high capacity automated production lines resulting in high quality value priced products. Ventilation ribs reduce condensation and allow air circulation.Specifications Consistent flatness allows for even media distribution Stackable – Lids contain stacking rings for increased stability Stackable/Slideable – Features partial edge ring on base which allows a single dish to be slid from a large stack and enhances stability when stacked Slippable – Designed for automated filling and streaking systems features side arrows and ISO Mark Target for automation Working Volume 1516mL Growth Area 55cm2 Case Quantity 500
Catalog No. 50-233-0346 Supplier Bellco Glass, Inc. Supplier No. 229694000
CELLTREAT Petri Dishes are manufactured in high capacity automated production lines resulting in high quality value priced products. Ventilation ribs reduce condensation and allow air circulation.Specifications Consistent flatness allows for even media distribution Stackable – Lids contain stacking rings for increased stability Stackable/Slideable – Features partial edge ring on base which allows a single dish to be slid from a large stack and enhances stability when stacked Slippable – Designed for automated filling and streaking systems features side arrows and ISO Mark Target for automation Working Volume 1516mL Growth Area 55cm2 Case Quantity 500