Learn More
Bellco Glass, Inc. 100mm x 15mm Tissue Culture
SDP
Catalog No. 502330325
Encompass_Preferred
Cell Culture Dishes are available in Tissue Culture Treated and Nontreated Cell Culture versions.  60 70 and 100mm dishes available with a pronounced grip ring for improved handling.Specifications Vented lids contain molded spacers to allow gas exchange except Part No. 229655000 which has a nonvented lid Working Volume 1516mL Working Area 55cm2 Case Quantity 500
Catalog No. 50-233-0325 Supplier Bellco Glass, Inc. Supplier No. 229690000
Cell Culture Dishes are available in Tissue Culture Treated and Nontreated Cell Culture versions.  60 70 and 100mm dishes available with a pronounced grip ring for improved handling.Specifications Vented lids contain molded spacers to allow gas exchange except Part No. 229655000 which has a nonvented lid Working Volume 1516mL Working Area 55cm2 Case Quantity 500